ContactUsLinkComponent
Radioactivity indicator dots, SP Bel-Art
Radioactivity indicator dots, SP Bel-Art
Catalog # 116-1095
undefined
Radioactivity indicator dots, SP Bel-Art
Catalog # 116-1095
Specifications
  • Colour:
    Red/yellow
  • Description:
    Indicator dots
  • Dimensions:
    6 mm
  • Packed:
    1000 labels/roll
  • Pk:
    1 Roll
Specifications
About this item
Stick-on dots with the radiation symbol on yellow background, which enables easy labelling of radioactive vessels.
  • Ideal for marking micro-centrifuge tubes
Colour coding complies with OSHA regulations.