Orders LinkContactUsLinkComponent
Label dots
Label dots
Certificates
Label dots
8 Options Available Below
Product Details & Documents
Self adhesive, smudge proof labels. Suitable for labelling samples and documents. Bright colours. Accepts a variety of inks.
  • Oil and water-resistant
  • Temperature resistant from −45 to +121 °C
  • Peels off easily and leaves no residue
Diameter: 19,0 mm
Product Family Options
Product Information
  • Material
    Colour
    Dimensions
    Availability
    Price
  • 817-1652
    Label dots
    Label dots
    Paper
    Pale purple
    19 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 52,10 €
    52,10 €
    1 * 1.000 ST
       52,10 €
  • 817-1647
    Label dots
    Label dots
    Paper
    Yellow
    19 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 52,10 €
    52,10 €
    1 * 1.000 ST
       52,10 €
  • 817-1648
    Label dots
    Label dots
    Paper
    Red
    19 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 52,10 €
    52,10 €
    1 * 1.000 ST
       52,10 €
  • 817-1649
    Label dots
    Label dots
    Paper
    Blue
    19 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 52,10 €
    52,10 €
    1 * 1.000 ST
       52,10 €
  • 817-1653
    Label dots
    Label dots
    Paper
    Dark blue
    19 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 52,10 €
    52,10 €
    1 * 1.000 ST
       52,10 €
  • 817-0378
    Fluorescent label dots
    Fluorescent label dots
    Paper
    Pink
    19 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 54,80 €
    54,80 €
    1 * 1.000 ST
       54,80 €
  • 817-0379
    Fluorescent label dots
    Fluorescent label dots
    Paper
    Green
    19 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 54,80 €
    54,80 €
    1 * 1.000 ST
       54,80 €
  • 817-0380
    Fluorescent label dots
    Fluorescent label dots
    Paper
    Orange
    19 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 54,80 €
    54,80 €
    1 * 1.000 ST
       54,80 €