ContactUsLinkComponent
Staining jar according to Coplin
Staining jar according to Coplin
Catalog # 720-0576
Supplier:  SMITH SCIENTIFIC
undefined
Staining jar according to Coplin
Catalog # 720-0576
Supplier:  SMITH SCIENTIFIC
Specifications
  • Description:
    Staining jar acc. to Coplin
  • Pk:
    1
Specifications
About this item
Soda-lime glass. With grooves, to accept ten 76×26 mm slides, vertically, back to back.
  • Complete with drop-on lid