Orders LinkContactUsLinkComponent
Staining systems, EasyDip™
Staining systems, EasyDip™
Supplier:   Simport Scientific
Staining systems, EasyDip™
Supplier:  Simport Scientific
Certificates
Staining systems, EasyDip™
Staining systems, EasyDip™
9 Options Available Below
Product Details & Documents
The slide staining system is made of acetal plastic.
  • Jars can be loosely joined to each other to keep the same order when moved around
  • Wide base for stability while inside is recessed to reduce reagent volume
  • Available in five colours to aid identification of contents or applications
The slide staining system has two components: A square staining jar and a 12 position vertical slide rack. The staining jar is made of resistant acetal plastic and will resist most staining agents including alcohol and xylene but not phenol. Autoclavable, it is therefore suitable for special stains, frozen sections and special processes.
Volume: 80 ml
W×D×H: 76×64×92 mm
Ordering information: The staining jars and staining rack are sold separately, except for the assorted package (720-0791), which contains one jar of each colour (blue, green, pink, white, yellow) and a rack. The EasyDip aluminum holder is designed to hold six EasyDip slide staining jars. The EasyDip slide staining kit (SIMPM906-12AS) contains 6 jars (blue, green, pink, 2xwhite, yellow), an anodized aluminium rack, and one slide staining rack.
Product Family Options
Product Information
  • Colour
    Material
    W×D×H
    Availability
    Price
  • 720-0791
    Slide staining rack and five jars
    Slide staining rack and five jars
    Assorted
    Acetal
    76×64×92 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 216,00 €
    216,00 €
    1 * 1 KIT
       216,00 €
  • 720-0792
    Slide staining cuvettes
    Slide staining cuvettes
    Blue
    Acetal
    76×64×92 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 176,00 €
    176,00 €
    1 * 6 ST
       176,00 €
  • 720-0794
    Slide staining cuvettes
    Slide staining cuvettes
    Green
    Acetal
    76×64×92 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 176,00 €
    176,00 €
    1 * 6 ST
       176,00 €
  • 720-0795
    Slide staining cuvettes
    Slide staining cuvettes
    Rose
    Acetal
    76×64×92 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 176,00 €
    176,00 €
    1 * 6 ST
       176,00 €
  • 720-0796
    Slide staining cuvettes
    Slide staining cuvettes
    White
    Acetal
    76×64×92 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 176,00 €
    176,00 €
    1 * 6 ST
       176,00 €
  • 720-0797
    Slide staining cuvettes
    Slide staining cuvettes
    Yellow
    Acetal
    76×64×92 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 176,00 €
    176,00 €
    1 * 6 ST
       176,00 €
  • 720-0793
    Vertical slide staining racks
    Vertical slide staining racks
    Dark grey
    Acetal
    76×64×92 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 194,00 €
    194,00 €
    1 * 6 ST
       194,00 €
  • SIMPM906-12AS
    EasyDip™ slide staining kit
    EasyDip™ slide staining kit
    Assorted
    Acetal
    76×64×92 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 346,00 €
    346,00 €
    1 * 1 ST
       346,00 €
Holder for EasyDip
Product Information
  • Colour
    Material
    W×D×H
    Availability
    Price
  • 630-1540
    Holder (for up to six jars)
    Holder (for up to six jars)
    Stainless steel
    425×102×38 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 195,00 €
    195,00 €
    1 * 1 ST
       195,00 €