Staining jars
Staining jar with 2 lids, 81×101×70 mm
Artikelnr. 631-9511
Anbieter: KARTELL
Jump to:
Product Details & Documents
PMP.
- One lid for when rack is not in use, providing evaporation free storage of staining liquid
- Second lid with special opening to allow handle of staining rack (631-9520) to be inserted
Staining jar supplied with two lids.
Delivery information: For use with 631-9520.
Caution: Xylene can cause etching after two days at room temperature.
Specifications
- Description:Staining jar with 2 lids
- W×D×H:81×101×70 mm