ContactUsLinkComponent
Brine tester
Brine tester
Catalog # 641-1500
Supplier:  Alla France
undefined
Brine tester
Catalog # 641-1500
Supplier:  Alla France
Specifications
  • Description:
    Brinometer
  • Length:
    290 mm
  • Pk:
    1
  • Range:
    0 – 26 %
Specifications
About this item
Brine is the percentage of saturated solution of salt, usually sodium chloride, in water.
A brinometer is used to measure the percentage of saturated solution of brine.