Order Entry
Austria
ContactUsLinkComponent
Milk hydrometer with thermometer
Milk hydrometer with thermometer
  614-1501
undefined
Milk hydrometer with thermometer
  614-1501

 

  • Temperature Range:
    0...40  °C
  • Description:
    Dornic milk hydrometer with thermometer
  • Length:
    220 mm
  • Pk:
    1
  • Range:
    1,018 - 1,038 g/ml

 

 

This Dornic milk hydrometer with built-in thermometer is graduated from 18 to 38 (1018 to 1038).

  • Internal thermometer and temperature correction indicator Control thermometers available

Note: Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.