Orders LinkContactUsLinkComponent
Short hydrometers
Supplier:   Alla France
Short hydrometers
Supplier:  Alla France
Certificates
Short hydrometers
Short hydrometers
19 Options Available Below
Product Details & Documents
General purpose hydrometers calibrated at 20 °C.
  • Reading below the meniscus
Length 160 mm
Product Family Options
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • 0,700 - 0,760 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,50 €
    32,50 €
    1 * 1 ST
       32,50 €
  • 0,760 - 0,820 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,50 €
    22,50 €
    1 * 1 ST
       22,50 €
  • 0,820 - 0,880 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,50 €
    32,50 €
    1 * 1 ST
       32,50 €
  • 0,880 - 0,940 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,50 €
    32,50 €
    1 * 1 ST
       32,50 €
  • 0,940 - 1,000 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,50 €
    32,50 €
    1 * 1 ST
       32,50 €
  • 1,000 - 1,060 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,20 €
    26,20 €
    1 * 1 ST
       26,20 €
  • 1,060 - 1,120 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,20 €
    26,20 €
    1 * 1 ST
       26,20 €
  • 1,120 - 1,180 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,20 €
    26,20 €
    1 * 1 ST
       26,20 €
  • 1,180 - 1,240 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,20 €
    26,20 €
    1 * 1 ST
       26,20 €
  • 1,240 - 1,300 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,10 €
    38,10 €
    1 * 1 ST
       38,10 €
  • 1,300 - 1,360 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,50 €
    28,50 €
    1 * 1 ST
       28,50 €
  • 1,360 - 1,420 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,50 €
    28,50 €
    1 * 1 ST
       28,50 €
  • 1,420 - 1,480 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,50 €
    28,50 €
    1 * 1 ST
       28,50 €
  • 1,480 - 1,540 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,50 €
    28,50 €
    1 * 1 ST
       28,50 €
  • 1,540 - 1,600 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,50 €
    28,50 €
    1 * 1 ST
       28,50 €
  • 1,600 - 1,660 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,30 €
    41,30 €
    1 * 1 ST
       41,30 €
  • 1,660 - 1,720 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,30 €
    41,30 €
    1 * 1 ST
       41,30 €
  • 1,720 - 1,780 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 98,00 €
    98,00 €
    1 * 1 ST
       98,00 €
  • 1,780 - 1,840 g/ml
    0,001 g/ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 98,00 €
    98,00 €
    1 * 1 ST
       98,00 €