Orders LinkContactUsLinkComponent
Tissue grinders, Dounce type
Tissue grinders, Dounce type
Supplier:   DWK Life Sciences
Tissue grinders, Dounce type
Supplier:  DWK Life Sciences
Certificates
Tissue grinders, Dounce type
Tissue grinders, Dounce type
2 Options Available Below
Product Details & Documents
Wheaton Dura-Grind® stainless steel Dounce tissue grinders provide a rugged alternative to glass Dounce tissue grinders. Smooth pestle action requires less effort than glass tissue grinders. Precision machined  No. 316 stainless steel to low tolerance of 0,0124 mm.
  • Mortar has flat bottom to stand upright
  • Single pestle and mortar are supplied as a matched set
  • Not autoclavable
Weight: 0,9 kg
Product Family Options
Product Information
  • Capacity
    Ø ext.×length
    Total length
    Availability
    Price
  • 15 ml
    15,5×114 mm
    171 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 390,00 €
    1 390,00 €
    1 * 1 ST
       1 390,00 €
  • 40 ml
    25,5×114 mm
    171 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 740,00 €
    1 740,00 €
    1 * 1 ST
       1 740,00 €