Orders LinkContactUsLinkComponent
Tissue grinders, Dounce type
Tissue grinders, Dounce type
Supplier:   DWK Life Sciences
Tissue grinders, Dounce type
Supplier:  DWK Life Sciences
Certificates
Tissue grinders, Dounce type
Tissue grinders, Dounce type
4 Options Available Below
Product Details & Documents
Borosilicate glass. Designed to retain a high percentage of cell nuclei and mitochondria in soft tissues. In operation, the pestle ball is encircled in liquid which avoids heat build up. Friction is reduced and there is reduced heat accumulation.
  • Mortar has large reservoir and pouring lip
Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Certifications: Wheaton 33 low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
Delivery information: Each mortar is supplied with a 'loose' pestle (0,114 mm ±0,025 mm), and a 'tight' pestle (0,05 mm  ±0,025 mm).
Product Family Options
Product Information
  • Capacity
    Ø ext.×length
    Total length
    Availability
    Price
  • 1 ml
    11×48 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 483,00 €
    483,00 €
    1 * 2 ST
       483,00 €
  • 7 ml
    13×82 mm
    175 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 529,00 €
    529,00 €
    1 * 2 ST
       529,00 €
  • 15 ml
    22×94 mm
    210 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 608,00 €
    608,00 €
    1 * 2 ST
       608,00 €
  • 40 ml
    32×140 mm
    285 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 767,00 €
    767,00 €
    1 * 2 ST
       767,00 €