Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Histosette® I, M498

Tissue processing/embedding cassettes, Histosette® I, M498

Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M498
Supplier:  Simport Scientific
Certificates
Tissue processing/embedding cassettes, Histosette® I, M498
Tissue processing/embedding cassettes, Histosette® I, M498

11 Options Available Below

Product Details & Documents

Acetal polymer.

  • Resistant to histological solvents
  • Flow-through slots maximise fluid exchange and ensure proper drainage
  • Integral lid
  • 45° angle writing area to enable use with some automated cassette-labelling instruments
Product Family Options

Product Information

  • Colour
    Availability
    Price
  • White
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 229,00 €
    229,00 €
    1 * 1.500 ST
       229,00 €

  • Green
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 229,00 €
    229,00 €
    1 * 1.500 ST
       229,00 €

  • Salmon
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 229,00 €
    229,00 €
    1 * 1.500 ST
       229,00 €

  • Turquoise
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 229,00 €
    229,00 €
    1 * 1.500 ST
       229,00 €

  • Purple
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 229,00 €
    229,00 €
    1 * 1.500 ST
       229,00 €

  • Orange
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 229,00 €
    229,00 €
    1 * 1.500 ST
       229,00 €

  • Pink
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 229,00 €
    229,00 €
    1 * 1.500 ST
       229,00 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 229,00 €
    229,00 €
    1 * 1.500 ST
       229,00 €

  • Blue
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 229,00 €
    229,00 €
    1 * 1.500 ST
       229,00 €

  • Beige
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 229,00 €
    229,00 €
    1 * 1.500 ST
       229,00 €

  • Grey
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 229,00 €
    229,00 €
    1 * 1.500 ST
       229,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.