Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Micromesh™ M507
Biopsy processing/embedding cassettes, Micromesh™ M507
Biopsy processing/embedding cassettes, Micromesh™ M507
Supplier:  Simport Scientific
Certificates
Biopsy processing/embedding cassettes, Micromesh™ M507
Biopsy processing/embedding cassettes, Micromesh™ M507
11 Options Available Below
Product Details & Documents
Acetal polymer. Biopsy processing/embedding cassettes, Micromesh™ M507.
  • No need for biopsy pads
  • Large square compartment, 27 mm², suitable for needle biopsies
  • 1676 square openings (0,38 mm²) allow improved fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Documents
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • Purple
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 518,00 €
    518,00 €
    1 * 1.000 ST
       518,00 €
  • Orange
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 518,00 €
    518,00 €
    1 * 1.000 ST
       518,00 €
  • Turquoise
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 518,00 €
    518,00 €
    1 * 1.000 ST
       518,00 €
  • White
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 518,00 €
    518,00 €
    1 * 1.000 ST
       518,00 €
  • Pink
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 518,00 €
    518,00 €
    1 * 1.000 ST
       518,00 €
  • Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 518,00 €
    518,00 €
    1 * 1.000 ST
       518,00 €
  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 518,00 €
    518,00 €
    1 * 1.000 ST
       518,00 €
  • Blue
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 518,00 €
    518,00 €
    1 * 1.000 ST
       518,00 €
  • Salmon
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 518,00 €
    518,00 €
    1 * 1.000 ST
       518,00 €
  • Beige
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 518,00 €
    518,00 €
    1 * 1.000 ST
       518,00 €
  • Grey
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 518,00 €
    518,00 €
    1 * 1.000 ST
       518,00 €