Order Entry
Austria
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Histosette® I, M490

Tissue processing/embedding cassettes, Histosette® I, M490

Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Histosette® I, M490
Supplier:  Simport Scientific
Certificates
Tissue processing/embedding cassettes, Histosette® I, M490
Tissue processing/embedding cassettes, Histosette® I, M490

11 Options Available Below

Product Details & Documents

Acetal polymer.

  • Resistant to histological solvents
  • Flow-through slots maximise fluid exchange and ensure proper drainage
  • Integral lid eliminates the need for separate steel lids
  • 30° writing angle
Product Family Options

Product Information

  • Colour
    Availability
    Price
  • Purple
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 163,00 €
    163,00 €
    1 * 1.500 ST
       163,00 €

  • Turquoise
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 163,00 €
    163,00 €
    1 * 1.500 ST
       163,00 €

  • White
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 163,00 €
    163,00 €
    1 * 1.500 ST
       163,00 €

  • Pink
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 163,00 €
    163,00 €
    1 * 1.500 ST
       163,00 €

  • Green
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 126,00 €
    126,00 €
    1 * 1.500 ST
       126,00 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 163,00 €
    163,00 €
    1 * 1.500 ST
       163,00 €

  • Blue
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 163,00 €
    163,00 €
    1 * 1.500 ST
       163,00 €

  • Salmon
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 163,00 €
    163,00 €
    1 * 1.500 ST
       163,00 €

  • Grey
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 163,00 €
    163,00 €
    1 * 1.500 ST
       163,00 €

  • Orange
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 126,00 €
    126,00 €
    1 * 1.500 ST
       126,00 €

  • Beige
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 163,00 €
    163,00 €
    1 * 1.500 ST
       163,00 €