Orders LinkContactUsLinkComponent
Goggles, Elite
Goggles, Elite
Anti-UV, anti-fog, Clear acetate, Transparent
Catalog # 111-0302
Supplier:   Bollé Safety
Goggles, Elite
Catalog # 111-0302
Supplier:  Bollé Safety
Goggles, Elite
Goggles, Elite
Anti-UV, anti-fog, Clear acetate, Transparent
Price
25,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Comfortable goggles which can be worn with prescription eyewear.
  • Extra-wide panoramic vision
  • Indirect ventilation
  • Pivoting, adjustable headband
Weight: 121 g
EN 166, EN 170
Certifications: EN 166, EN 170
Specifications
  • Type:
    Anti-UV, anti-fog
  • Colour:
    Transparent
  • Lenses:
    Clear acetate