Orders LinkContactUsLinkComponent
Goggles, coverall
Anti-fog, anti-scratch, Clear PC, Translucent
Catalog # BOLECOVACLEAN
Supplier:   Bollé Safety
Goggles, coverall
Catalog # BOLECOVACLEAN
Supplier:  Bollé Safety
Goggles, coverall
Goggles, coverall
Anti-fog, anti-scratch, Clear PC, Translucent
Price
11,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Comfortable goggles with adjustable elastic straps.
  • Fit comfortably over most prescription eyewear
  • High flow ventilation with four indirect ventilators
  • Resistant to molten metal and hot solids, dust and liquid aerosols
 : EN 166
Specifications
  • Type:
    Anti-fog, anti-scratch
  • Colour:
    Translucent
  • Seq:
    01
  • Lenses:
    Clear PC