Orders LinkContactUsLinkComponent
Cryo-Gloves® Cold Resistant Gloves
Cryo-Gloves® Cold Resistant Gloves
Supplier:   TEMPSHIELD
Cryo-Gloves® Cold Resistant Gloves
Supplier:  TEMPSHIELD
Certificates
Cryo-Gloves® Cold Resistant Gloves
33 Options Available Below
Product Details & Documents
These hand-specific gloves protect hands and arms against the hazards of working in and with ultra-cold temperatures down to –196 °C. Lightweight, breathable, and comfortable, they provide protection from liquid splashes and cryogenic temperatures but are not safe for immersion in cryogenic liquids.
  • Provide warmth while retaining flexibility even after prolonged exposure to cryogenic atmospheres
  • Cryo-Gloves® allow sufficient dexterity to handle small objects and controls
  • Inner cuffs (except for the wrist length models) contain a single layer of insulation material, a scrim backing and a nylon taffeta lining
  • Wrist length models feature a knitted cuff, all other models feature a gauntlet cuff
  • Sewn construction
Cryo-Gloves® are offered in three configurations:
Water resistant: These gloves comprise of an outer shell consisting of nylon Taslan® with a liquid-proof, breathable membrane on the inner surface, and an inner insulated glove consisting of three layers (non woven synthetic insulation material + lining material).
Waterproof: These gloves have the same construction as the water-resistant gloves but additionally feature a seamless, breathable, waterproof lining.
Industrial: A rugged, abrasion resistant outer shell and a thin, seamless 100% waterproof liner provide extra protection from cryogenic splashes and an excellent grip for heavy-duty applications.
EN 388, EN 511, Cat. III
Certifications: EN 388, EN 511, Cat. III
Specifications for Product Family
Specifications
  • Catalog No:
  • 112-2722
  • 112-2720
  • 112-2718
  • 112-9002
  • 112-2709
  • 112-2707
  • 112-0343
  • 112-2710
  • 112-0528
  • 112-2702
  • 112-2700
  • 112-2704
  • 112-0530
  • 112-2714
  • 112-2712
  • 112-2716
  • 112-2723
  • 112-2721
  • 112-2719
  • 112-2724
  • TEMPMAXXLWP
  • 112-9001
  • 112-2708
  • 112-2706
  • 112-2711
  • 112-0529
  • 112-2703
  • 112-2701
  • 112-9000
  • 112-0531
  • 112-2715
  • 112-2713
  • 112-2717
  • Model:
  • Wrist
  • Wrist
  • Wrist
  • Wrist
  • Mid-arm
  • Mid-arm
  • Mid-arm
  • Mid-arm
  • Elbow
  • Elbow
  • Elbow
  • Elbow
  • Shoulder
  • Shoulder
  • Shoulder
  • Shoulder
  • Wrist
  • Wrist
  • Wrist
  • Wrist
  • Mid-arm
  • Mid-arm
  • Mid-arm
  • Mid-arm
  • Mid-arm
  • Elbow
  • Elbow
  • Elbow
  • Elbow
  • Shoulder
  • Shoulder
  • Shoulder
  • Shoulder
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • EN norm:
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (32X)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
  • EN 388 (1323), EN 511 (321)
Specifications
Product Family Options
Water resistant
Product Information
  • Model
    Glove size
    Colour
    Length
    Availability
    Price
  • Wrist
    S
    Blue
    290 - 325 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 189,00 €
    189,00 €
    1 * 1 PAAR
       189,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2722
    Model
    Wrist
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Wrist
    M
    Blue
    290 - 325 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 189,00 €
    189,00 €
    1 * 1 PAAR
       189,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2720
    Model
    Wrist
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Wrist
    L
    Blue
    290 - 325 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 189,00 €
    189,00 €
    1 * 1 PAAR
       189,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2718
    Model
    Wrist
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Wrist
    XL
    Blue
    290 - 325 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 189,00 €
    189,00 €
    1 * 1 PAAR
       189,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-9002
    Model
    Wrist
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Mid-arm
    S
    Blue
    345 - 390 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 202,00 €
    202,00 €
    1 * 1 PAAR
       202,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2709
    Model
    Mid-arm
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Mid-arm
    M
    Blue
    345 - 390 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 202,00 €
    202,00 €
    1 * 1 PAAR
       202,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2707
    Model
    Mid-arm
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Mid-arm
    L
    Blue
    345 - 390 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 202,00 €
    202,00 €
    1 * 1 PAAR
       202,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-0343
    Model
    Mid-arm
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Mid-arm
    XL
    Blue
    345 - 390 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 202,00 €
    202,00 €
    1 * 1 PAAR
       202,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2710
    Model
    Mid-arm
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Elbow
    S
    Blue
    440 - 485 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 251,00 €
    251,00 €
    1 * 1 PAAR
       251,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-0528
    Model
    Elbow
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Elbow
    M
    Blue
    440 - 485 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 251,00 €
    251,00 €
    1 * 1 PAAR
       251,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2702
    Model
    Elbow
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Elbow
    L
    Blue
    440 - 485 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 251,00 €
    251,00 €
    1 * 1 PAAR
       251,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2700
    Model
    Elbow
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Elbow
    XL
    Blue
    440 - 485 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 251,00 €
    251,00 €
    1 * 1 PAAR
       251,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2704
    Model
    Elbow
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Shoulder
    S
    Blue
    625 - 685 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 332,00 €
    332,00 €
    1 * 1 PAAR
       332,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-0530
    Model
    Shoulder
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Shoulder
    M
    Blue
    625 - 685 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 332,00 €
    332,00 €
    1 * 1 PAAR
       332,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2714
    Model
    Shoulder
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Shoulder
    L
    Blue
    625 - 685 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 332,00 €
    332,00 €
    1 * 1 PAAR
       332,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2712
    Model
    Shoulder
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
  • Shoulder
    XL
    Blue
    625 - 685 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 332,00 €
    332,00 €
    1 * 1 PAAR
       332,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2716
    Model
    Shoulder
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (32X)
Waterproof
Product Information
  • Model
    Glove size
    Colour
    Length
    Availability
    Price
  • Wrist
    S
    Blue
    290 - 325 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 231,00 €
    231,00 €
    1 * 1 PAAR
       231,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2723
    Model
    Wrist
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Wrist
    M
    Blue
    290 - 325 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 231,00 €
    231,00 €
    1 * 1 PAAR
       231,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2721
    Model
    Wrist
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Wrist
    L
    Blue
    290 - 325 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 231,00 €
    231,00 €
    1 * 1 PAAR
       231,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2719
    Model
    Wrist
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Wrist
    XL
    Blue
    290 - 325 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 231,00 €
    231,00 €
    1 * 1 PAAR
       231,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2724
    Model
    Wrist
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Mid-arm
    XXL
    Blue
    335 - 395 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 279,00 €
    279,00 €
    1 * 1 PAAR
       279,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    TEMPMAXXLWP
    Model
    Mid-arm
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Mid-arm
    S
    Blue
    345 - 390 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 249,00 €
    249,00 €
    1 * 1 PAAR
       249,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-9001
    Model
    Mid-arm
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Mid-arm
    M
    Blue
    345 - 390 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 249,00 €
    249,00 €
    1 * 1 PAAR
       249,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2708
    Model
    Mid-arm
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Mid-arm
    L
    Blue
    345 - 390 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 249,00 €
    249,00 €
    1 * 1 PAAR
       249,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2706
    Model
    Mid-arm
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Mid-arm
    XL
    Blue
    345 - 390 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 249,00 €
    249,00 €
    1 * 1 PAAR
       249,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2711
    Model
    Mid-arm
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Elbow
    S
    Blue
    440 - 485 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 307,00 €
    307,00 €
    1 * 1 PAAR
       307,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-0529
    Model
    Elbow
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Elbow
    M
    Blue
    440 - 485 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 307,00 €
    307,00 €
    1 * 1 PAAR
       307,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2703
    Model
    Elbow
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Elbow
    L
    Blue
    440 - 485 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 307,00 €
    307,00 €
    1 * 1 PAAR
       307,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2701
    Model
    Elbow
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Elbow
    XL
    Blue
    440 - 485 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 307,00 €
    307,00 €
    1 * 1 PAAR
       307,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-9000
    Model
    Elbow
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Shoulder
    S
    Blue
    625 - 685 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 392,00 €
    392,00 €
    1 * 1 PAAR
       392,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-0531
    Model
    Shoulder
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Shoulder
    M
    Blue
    625 - 685 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 392,00 €
    392,00 €
    1 * 1 PAAR
       392,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2715
    Model
    Shoulder
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Shoulder
    L
    Blue
    625 - 685 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 392,00 €
    392,00 €
    1 * 1 PAAR
       392,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2713
    Model
    Shoulder
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)
  • Shoulder
    XL
    Blue
    625 - 685 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 392,00 €
    392,00 €
    1 * 1 PAAR
       392,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-2717
    Model
    Shoulder
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    EN norm
    EN 388 (1323), EN 511 (321)