Orders LinkContactUsLinkComponent
Glove liners, polyester, BCR™
Glove liners, polyester, BCR™
Glove liners, polyester, BCR™
Supplier:  BERKSHIRE
Certificates
Glove liners, polyester, BCR™
4 Options Available Below
Product Details & Documents
Continuous-filament knitted, these ambidextrous polyester gloves provide protection and comfort during extended wear. Ideal under latex, PVC, or other outer gloves to absorb perspiration from hands during periods of extended wear.
  • Seamless construction prevents unravelling and release of unwanted particles into critical environments
  • Launderable for repeated use
  • Comfortable half-finger design provides greater dexterity than a full-finger glove liner and has reduced courses in the finger and cuff
  • Full-finger style provides complete protection from irritation caused by latex and PVC gloves
Specifications for Product Family
Specifications
  • Catalog No:
  • 115-0537
  • 112-0616
  • 112-0532
  • 112-3344
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
Specifications
Product Family Options
Product Information
  • Model
    Glove size
    Colour
    Packed
    Availability
    Price
  • Half-finger liners
    M
    White
    200 pairs/pack (bulk)
    Log in for availability
    Log in for availability
    1 * 200 PAAR
    1 * 200 PAAR - 1 200,00 €
    1 200,00 €
    1 * 200 PAAR
       1 200,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    115-0537
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
  • Half-finger liners
    L
    White
    10 pairs/pack
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 31,90 €
    31,90 €
    1 * 10 PAAR
       31,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-0616
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
  • Full-finger liners
    M
    White
    10 pairs/pack
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 68,10 €
    68,10 €
    1 * 10 PAAR
       68,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-0532
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
  • Full-finger liners
    L
    White
    10 pairs/pack
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 55,50 €
    55,50 €
    1 * 10 PAAR
       55,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-3344
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable