Orders LinkContactUsLinkComponent
Separating funnels, pear shaped
Supplier:   SciLabware
Separating funnels, pear shaped
Supplier:  SciLabware
Certificates
Separating funnels, pear shaped
Separating funnels, pear shaped
7 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Available with ground glass stopper and interchangeable PTFE key.
Product Family Options
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 50 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,70 €
    72,70 €
    1 * 1 ST
       72,70 €
  • 100 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €
  • 250 ml
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,00 €
    86,00 €
    1 * 1 ST
       86,00 €
  • 500 ml
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 111,00 €
    111,00 €
    1 * 1 ST
       111,00 €
  • 1000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €
  • 2000 ml
    34/35 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 215,00 €
    215,00 €
    1 * 1 ST
       215,00 €
  • 5000 ml
    34/35 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 338,00 €
    338,00 €
    1 * 1 ST
       338,00 €