Order Entry
Austria
Orders LinkContactUsLinkComponent
Separating funnel holder

Separating funnel holder

for 1 separating funnel up from 75 mm Ø
Catalog # 527-1999
Supplier:   KARTELL
Separating funnel holder
Catalog # 527-1999
Supplier:  KARTELL
Separating funnel holder
Separating funnel holder

for 1 separating funnel up from 75 mm Ø

Price
13,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

PP.

Clamp fits rods from Ø 8 to 14 mm.

Not usable with funnels Ø less 75 mm.

Specifications
  • Description:
    for 1 separating funnel up from 75 mm Ø
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.