Orders LinkContactUsLinkComponent
Separating funnels, Squibb, Nalgene®
Separating funnels, Squibb, Nalgene®
   Sustainable Options Available
Separating funnels, Squibb, Nalgene®
   Sustainable Options Available
Certificates
Separating funnels, Squibb, Nalgene®
5 Options Available Below
Product Details & Documents
FEP translucent, TFE stopcock, ETFE screw closure.
  • Leakproof closure and stopcock
  • Autoclavable except for stopcock assembly
  • TFE stopcock requires no lubrication
Robust, transparent, non stick for easy cleaning, non wetting for complete draining and impervious to any chemical used in a separating funnel.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 527-2101
  • 527-2102
  • 527-2103
  • 527-2104
  • 527-2105
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Ø cap
    Availability
    Price
  • 125 ml
    28 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,00 €
    196,00 €
    1 * 1 ST
       196,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    527-2101
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 250 ml
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 218,00 €
    218,00 €
    1 * 1 ST
       218,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    527-2102
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 500 ml
    43 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 320,00 €
    320,00 €
    1 * 1 ST
       320,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    527-2103
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 1000 ml
    53 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 376,00 €
    376,00 €
    1 * 1 ST
       376,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    527-2104
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 2000 ml
    53 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 586,00 €
    586,00 €
    1 * 1 ST
       586,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    527-2105
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Replacement stopcock assembly
    Replacement stopcock assembly
    NALG71-4301-1050
    Log in for availability
    1 * 1 ST - 133,00 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.