Orders LinkContactUsLinkComponent
Separating funnels, Squibb, Nalgene®
   Sustainable Options Available
Separating funnels, Squibb, Nalgene®
   Sustainable Options Available
Certificates
Separating funnels, Squibb, Nalgene®
4 Options Available Below
Product Details & Documents
PPCO translucent, TFE stopcock, PP screw closure.
  • Leakproof closure and stopcock
  • Very good chemical resistance
  • Autoclavable except for stopcock assembly
  • TFE stopcock requires no lubrication
Specifications for Product Family
Specifications
  • Catalog No:
  • 527-0005
  • 527-0006
  • 527-0007
  • 527-0008
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Ø cap
    Availability
    Price
  • 125 ml
    28 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 100,00 €
    100,00 €
    1 * 1 ST
       100,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    527-0005
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 250 ml
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    527-0006
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 500 ml
    43 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 113,00 €
    113,00 €
    1 * 1 ST
       113,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    527-0007
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 1000 ml
    53 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 141,00 €
    141,00 €
    1 * 1 ST
       141,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    527-0008
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact