Orders LinkContactUsLinkComponent
Buchner funnels
Buchner funnels
Supplier:   DIXON SCIENTIFIC
Buchner funnels
Supplier:  DIXON SCIENTIFIC
Certificates
Buchner funnels
5 Options Available Below
Product Details & Documents
Borosilicate glass
  • With sintered glass disc
  • Very good chemical resistance
  • High temperature resistance
For paper or membrane filtration disc.
Product Family Options
Product Information
  • Plate Ø
    Capacity
    Porosity
    Availability
    Price
  • 30 mm
    35 ml
    P100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 88,60 €
    88,60 €
    1 * 1 ST
       88,60 €
  • 30 mm
    35 ml
    P40
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 95,00 €
    95,00 €
    1 * 1 ST
       95,00 €
  • 40 mm
    80 ml
    P100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 95,50 €
    95,50 €
    1 * 1 ST
       95,50 €
  • 40 mm
    80 ml
    P40
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 95,50 €
    95,50 €
    1 * 1 ST
       95,50 €
  • 60 mm
    140 ml
    P40
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 190,00 €
    190,00 €
    1 * 1 ST
       190,00 €