Orders LinkContactUsLinkComponent
Büchner funnels, Nalgene®
Büchner funnels, Nalgene®
   Sustainable Options Available
Büchner funnels, Nalgene®
   Sustainable Options Available
Certificates
Büchner funnels, Nalgene®
5 Options Available Below
Product Details & Documents
PP
  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning
  • Autoclavable
For filter paper or membrane filtration discs.
Specifications for Product Family
Specifications
  • Catalog No:
  • 221-0100
  • 221-0101
  • 221-0102
  • 221-0103
  • 221-0104
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Family Options
Product Information
  • Capacity
    For filter Ø
    Availability
    Price
  • 83 ml
    55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,40 €
    15,40 €
    1 * 1 ST
       15,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    221-0100
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 186 ml
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,00 €
    19,00 €
    1 * 1 ST
       19,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    221-0101
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 358 ml
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,10 €
    32,10 €
    1 * 1 ST
       32,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    221-0102
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 919 ml
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 46,50 €
    46,50 €
    1 * 1 ST
       46,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    221-0103
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 2906 ml
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,40 €
    58,40 €
    1 * 1 ST
       58,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    221-0104
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact