Orders LinkContactUsLinkComponent
Filter Funnels
Filter Funnels
Supplier:  ROBU GLASFILTER
Certificates
Filter Funnels
52 Options Available Below
Product Details & Documents
ROBU® Filter Funnel with VitraPOR® Filter is designed for high-performance filtration. Equipped with a VitraPOR® filter, it offers exceptional chemical resistance and precision, making it ideal for demanding laboratory and industrial filtration processes. The funnel is durable, reusable, and easy to clean.
  • VitraPOR® filter for superior performance
  • High chemical resistance
  • Precise filtration
  • Durable and reusable
  • Ideal for laboratory and industrial use
  • Easy to clean
  • Efficient filtration for demanding applications
  • Robust construction for long-lasting use
The ROBU Filter Funnel with VitraPOR filter is engineered for high-efficiency filtration in both laboratory and industrial settings. Featuring the VitraPOR filter, known for its exceptional chemical resistance and precision, this funnel delivers reliable performance in demanding filtration processes. Its robust construction ensures durability, while the funnel is easy to clean and can be reused, providing an environmentally friendly and cost-effective solution. Designed for use with various filter papers or membranes, it ensures optimal filtration, making it ideal for applications requiring precise and efficient result
 : In compliance with ISO 4796-80 and ASTM E128-99 standards.
Product Family Options
Product Information
  • Plate Ø
    Capacity
    Porosity
    Material
    Height
    Stem O-Ø
    Funnel O-Ø
    Availability
    Price
  • 511-0371
    Filter funnel
    Filter funnel
    36 mm
    50 ml
    0
    Borosilicate glass 3.3
    53 mm
    10 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,20 €
    38,20 €
    1 * 1 ST
       38,20 €
  • 511-1301
    Filter funnel
    Filter funnel
    36 mm
    50 ml
    1
    Borosilicate glass 3.3
    53 mm
    10 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,20 €
    38,20 €
    1 * 1 ST
       38,20 €
  • 511-1302
    Filter funnel
    Filter funnel
    36 mm
    50 ml
    2
    Borosilicate glass 3.3
    53 mm
    10 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,20 €
    38,20 €
    1 * 1 ST
       38,20 €
  • 511-1303
    Filter funnel
    Filter funnel
    36 mm
    50 ml
    3
    Borosilicate glass 3.3
    53 mm
    10 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,20 €
    38,20 €
    1 * 1 ST
       38,20 €
  • 511-1304
    Filter funnel
    Filter funnel
    36 mm
    50 ml
    4
    Borosilicate glass 3.3
    53 mm
    10 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,00 €
    40,00 €
    1 * 1 ST
       40,00 €
  • 511-1305
    Filter funnel
    Filter funnel
    36 mm
    50 ml
    5
    Borosilicate glass 3.3
    53 mm
    10 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,50 €
    59,50 €
    1 * 1 ST
       59,50 €
  • ROBU2150F
    Filter funnel
    Filter funnel
    36 mm
    50 ml
    Fine
    Borosilicate glass 3.3
    53 mm
    10 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,60 €
    45,60 €
    1 * 1 ST
       45,60 €
  • 511-0373
    Filter funnel
    Filter funnel
    48 mm
    75 ml
    0
    Borosilicate glass 3.3
    50 mm
    10 mm
    52 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,30 €
    41,30 €
    1 * 1 ST
       41,30 €
  • 511-1311
    Filter funnel
    Filter funnel
    48 mm
    75 ml
    1
    Borosilicate glass 3.3
    50 mm
    10 mm
    52 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,30 €
    41,30 €
    1 * 1 ST
       41,30 €
  • 511-1312
    Filter funnel
    Filter funnel
    48 mm
    75 ml
    2
    Borosilicate glass 3.3
    50 mm
    10 mm
    52 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,30 €
    41,30 €
    1 * 1 ST
       41,30 €
  • 511-1313
    Filter funnel
    Filter funnel
    48 mm
    75 ml
    3
    Borosilicate glass 3.3
    50 mm
    10 mm
    52 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,30 €
    41,30 €
    1 * 1 ST
       41,30 €
  • 511-1314
    Filter funnel
    Filter funnel
    48 mm
    75 ml
    4
    Borosilicate glass 3.3
    50 mm
    10 mm
    52 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,30 €
    44,30 €
    1 * 1 ST
       44,30 €
  • 511-1315
    Filter funnel
    Filter funnel
    48 mm
    75 ml
    5
    Borosilicate glass 3.3
    50 mm
    10 mm
    52 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,30 €
    64,30 €
    1 * 1 ST
       64,30 €
  • ROBU2175F
    Filter funnel
    Filter funnel
    48 mm
    75 ml
    Fine
    Borosilicate glass 3.3
    50 mm
    10 mm
    52 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,10 €
    50,10 €
    1 * 1 ST
       50,10 €
  • 511-0363
    Filter funnel
    Filter funnel
    60 mm
    125 ml
    0
    Borosilicate glass 3.3
    56 mm
    10 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,60 €
    51,60 €
    1 * 1 ST
       51,60 €
  • 511-1321
    Filter funnel
    Filter funnel
    60 mm
    125 ml
    1
    Borosilicate glass 3.3
    56 mm
    10 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,60 €
    51,60 €
    1 * 1 ST
       51,60 €
  • 511-1322
    Filter funnel
    Filter funnel
    60 mm
    125 ml
    2
    Borosilicate glass 3.3
    56 mm
    10 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,60 €
    51,60 €
    1 * 1 ST
       51,60 €
  • 511-1323
    Filter funnel
    Filter funnel
    60 mm
    125 ml
    3
    Borosilicate glass 3.3
    56 mm
    10 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,60 €
    51,60 €
    1 * 1 ST
       51,60 €
  • 511-1324
    Filter funnel
    Filter funnel
    60 mm
    125 ml
    4
    Borosilicate glass 3.3
    56 mm
    10 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,20 €
    56,20 €
    1 * 1 ST
       56,20 €
  • 511-1325
    Filter funnel
    Filter funnel
    60 mm
    125 ml
    5
    Borosilicate glass 3.3
    56 mm
    10 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,20 €
    92,20 €
    1 * 1 ST
       92,20 €
  • ROBU2112F
    Filter funnel
    Filter funnel
    60 mm
    125 ml
    Fine
    Borosilicate glass 3.3
    56 mm
    10 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,40 €
    65,40 €
    1 * 1 ST
       65,40 €
  • 511-0364
    Filter funnel
    Filter funnel
    80 mm
    250 ml
    0
    Borosilicate glass 3.3
    65 mm
    18 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,20 €
    71,20 €
    1 * 1 ST
       71,20 €
  • 511-0365
    Filter funnel
    Filter funnel
    80 mm
    250 ml
    1
    Borosilicate glass 3.3
    65 mm
    18 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,20 €
    71,20 €
    1 * 1 ST
       71,20 €
  • 511-0366
    Filter funnel
    Filter funnel
    80 mm
    250 ml
    2
    Borosilicate glass 3.3
    65 mm
    18 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,20 €
    71,20 €
    1 * 1 ST
       71,20 €
  • 511-0367
    Filter funnel
    Filter funnel
    80 mm
    250 ml
    3
    Borosilicate glass 3.3
    65 mm
    18 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 71,20 €
    71,20 €
    1 * 1 ST
       71,20 €
  • 511-0368
    Filter funnel
    Filter funnel
    80 mm
    250 ml
    4
    Borosilicate glass 3.3
    65 mm
    18 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 81,20 €
    81,20 €
    1 * 1 ST
       81,20 €
  • 511-0369
    Filter funnel
    Filter funnel
    80 mm
    250 ml
    5
    Borosilicate glass 3.3
    65 mm
    18 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 123,00 €
    123,00 €
    1 * 1 ST
       123,00 €
  • ROBU2125F
    Filter funnel
    Filter funnel
    80 mm
    250 ml
    Fine
    Borosilicate glass 3.3
    65 mm
    18 mm
    85 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,00 €
    94,00 €
    1 * 1 ST
       94,00 €
  • 511-0372
    Filter funnel
    Filter funnel
    95 mm
    500 ml
    0
    Borosilicate glass 3.3
    100 mm
    22 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,20 €
    86,20 €
    1 * 1 ST
       86,20 €
  • 511-1331
    Filter funnel
    Filter funnel
    95 mm
    500 ml
    1
    Borosilicate glass 3.3
    100 mm
    22 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,20 €
    86,20 €
    1 * 1 ST
       86,20 €
  • 511-1332
    Filter funnel
    Filter funnel
    95 mm
    500 ml
    2
    Borosilicate glass 3.3
    100 mm
    22 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,20 €
    86,20 €
    1 * 1 ST
       86,20 €
  • 511-1333
    Filter funnel
    Filter funnel
    95 mm
    500 ml
    3
    Borosilicate glass 3.3
    100 mm
    22 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,20 €
    86,20 €
    1 * 1 ST
       86,20 €
  • 511-1334
    Filter funnel
    Filter funnel
    95 mm
    500 ml
    4
    Borosilicate glass 3.3
    100 mm
    22 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €
  • 511-1335
    Filter funnel
    Filter funnel
    95 mm
    500 ml
    5
    Borosilicate glass 3.3
    100 mm
    22 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 179,00 €
    179,00 €
    1 * 1 ST
       179,00 €
  • ROBU2155F
    Filter funnel
    Filter funnel
    95 mm
    500 ml
    Fine
    Borosilicate glass 3.3
    100 mm
    22 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 118,00 €
    118,00 €
    1 * 1 ST
       118,00 €
  • 511-0362
    Filter funnel
    Filter funnel
    124 mm
    1000 ml
    0
    Borosilicate glass 3.3
    115 mm
    22 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 142,00 €
    142,00 €
    1 * 1 ST
       142,00 €
  • 511-1341
    Filter funnel
    Filter funnel
    124 mm
    1000 ml
    1
    Borosilicate glass 3.3
    115 mm
    22 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 142,00 €
    142,00 €
    1 * 1 ST
       142,00 €
  • 511-1342
    Filter funnel
    Filter funnel
    124 mm
    1000 ml
    2
    Borosilicate glass 3.3
    115 mm
    22 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 142,00 €
    142,00 €
    1 * 1 ST
       142,00 €
  • 511-1343
    Filter funnel
    Filter funnel
    124 mm
    1000 ml
    3
    Borosilicate glass 3.3
    115 mm
    22 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 142,00 €
    142,00 €
    1 * 1 ST
       142,00 €
  • 511-1344
    Filter funnel
    Filter funnel
    124 mm
    1000 ml
    4
    Borosilicate glass 3.3
    115 mm
    22 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 183,00 €
    183,00 €
    1 * 1 ST
       183,00 €
  • 511-1345
    Filter funnel
    Filter funnel
    124 mm
    1000 ml
    5
    Borosilicate glass 3.3
    115 mm
    22 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 307,00 €
    307,00 €
    1 * 1 ST
       307,00 €
  • ROBU2111F
    Filter funnel
    Filter funnel
    124 mm
    1000 ml
    Fine
    Borosilicate glass 3.3
    115 mm
    22 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 195,00 €
    195,00 €
    1 * 1 ST
       195,00 €
  • 511-0370
    Filter funnel
    Filter funnel
    180 mm
    4000 ml
    0
    Borosilicate glass 3.3
    206 mm
    30 mm
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • 511-1351
    Filter funnel
    Filter funnel
    180 mm
    4000 ml
    1
    Borosilicate glass 3.3
    206 mm
    30 mm
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • 511-1352
    Filter funnel
    Filter funnel
    180 mm
    4000 ml
    2
    Borosilicate glass 3.3
    206 mm
    30 mm
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • 511-1353
    Filter funnel
    Filter funnel
    180 mm
    4000 ml
    3
    Borosilicate glass 3.3
    206 mm
    30 mm
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • Filter funnel
    180 mm
    4000 ml
    4
    Borosilicate glass 3.3
    206 mm
    30 mm
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 483,00 €
    483,00 €
    1 * 1 ST
       483,00 €
  • 511-0374
    Filter funnel
    Filter funnel
    205 mm
    8000 ml
    0
    Borosilicate glass 3.3
    280 mm
    30 mm
    215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 780,00 €
    780,00 €
    1 * 1 ST
       780,00 €
  • 511-0375
    Filter funnel
    Filter funnel
    205 mm
    8000 ml
    1
    Borosilicate glass 3.3
    280 mm
    30 mm
    215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 780,00 €
    780,00 €
    1 * 1 ST
       780,00 €
  • 511-1316
    Filter funnel
    Filter funnel
    205 mm
    8000 ml
    2
    Borosilicate glass 3.3
    280 mm
    30 mm
    215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 780,00 €
    780,00 €
    1 * 1 ST
       780,00 €
  • 511-0376
    Filter funnel
    Filter funnel
    205 mm
    8000 ml
    3
    Borosilicate glass 3.3
    280 mm
    30 mm
    215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 780,00 €
    780,00 €
    1 * 1 ST
       780,00 €
  • 511-0377
    Filter funnel
    Filter funnel
    205 mm
    8000 ml
    4
    Borosilicate glass 3.3
    280 mm
    30 mm
    215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 898,00 €
    898,00 €
    1 * 1 ST
       898,00 €
Rubber gasket, GUKO
Product Information
  • Catalog #
    Ext.Ø
    Description
    Availability
    Price
  • 22 mm
    Rubber gasket, GUKO, inner Ø: 6 mm, size: 1
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 11,80 €
    11,80 €
    1 * 5 ST
       11,80 €
Product Information
  • Catalog #
    Ext.Ø
    Description
    Availability
    Price
  • 28 mm
    Rubber gasket, GUKO, inner Ø: 11 mm, size: 2
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 13,00 €
    13,00 €
    1 * 5 ST
       13,00 €
Product Information
  • Catalog #
    Ext.Ø
    Description
    Availability
    Price
  • 33 mm
    Rubber gasket, GUKO, inner Ø: 15 mm, size: 3
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 16,30 €
    16,30 €
    1 * 5 ST
       16,30 €
Product Information
  • Catalog #
    Ext.Ø
    Description
    Availability
    Price
  • 40 mm
    Rubber gasket, GUKO, inner Ø: 18 mm, size: 4
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 18,00 €
    18,00 €
    1 * 5 ST
       18,00 €
Product Information
  • Catalog #
    Ext.Ø
    Description
    Availability
    Price
  • 53 mm
    Rubber gasket, GUKO, inner Ø: 25 mm, size: 5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 21,60 €
    21,60 €
    1 * 5 ST
       21,60 €
Product Information
  • Catalog #
    Ext.Ø
    Description
    Availability
    Price
  • 67 mm
    Rubber gasket, GUKO, inner Ø: 32 mm, size: 6
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 24,30 €
    24,30 €
    1 * 5 ST
       24,30 €
Product Information
  • Catalog #
    Ext.Ø
    Description
    Availability
    Price
  • 77 mm
    Rubber gasket, GUKO, inner Ø: 45 mm, size: 7
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 27,30 €
    27,30 €
    1 * 5 ST
       27,30 €
Product Information
  • Catalog #
    Ext.Ø
    Description
    Availability
    Price
  • 88 mm
    Rubber gasket, GUKO, inner Ø: 52 mm, size: 8
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 36,00 €
    36,00 €
    1 * 5 ST
       36,00 €
Rubber gaskets, GUKO set
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Rubber gaskets, GUKO set, size: 1 - 5
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,20 €
    16,20 €
    1 * 1 ST
       16,20 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Rubber gaskets, GUKO set, size: 1 - 7
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,40 €
    26,40 €
    1 * 1 ST
       26,40 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Rubber gaskets, GUKO set, size: 1 - 8
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,60 €
    33,60 €
    1 * 1 ST
       33,60 €