Micro-Filter Funnels
About this item
ROBU® Micro-filter funnels with integrated VitraPOR® glass filters are ideal for precise filtration of very small liquid volumes in analytical and preparative laboratory processes.
For very small volumes
- Integrated VitraPOR® filter
- High chemical resistance
- Reusable and compact design
- Fine filtration
- Ideal for analytics and micro-preparations
The ROBU Micro-filter funnels are specially designed for filtration tasks involving minimal liquid quantities. Equipped with a high-performance VitraPOR glass filter, these funnels deliver reliable particle retention and reproducible filtration results.
Their compact size makes them ideal for analytical applications, research environments, and sample preparation where space and precision are key. Made from high-grade borosilicate glass, they offer excellent chemical resistance and are reusable for sustainable lab workflows.
: In compliance with ISO 4796-80 and ASTM E128-99 standards.
10 Options Available Below
Product Details & Documents
ROBU® Micro-filter funnels with integrated VitraPOR® glass filters are ideal for precise filtration of very small liquid volumes in analytical and preparative laboratory processes.
For very small volumes
- Integrated VitraPOR® filter
- High chemical resistance
- Reusable and compact design
- Fine filtration
- Ideal for analytics and micro-preparations
The ROBU Micro-filter funnels are specially designed for filtration tasks involving minimal liquid quantities. Equipped with a high-performance VitraPOR glass filter, these funnels deliver reliable particle retention and reproducible filtration results.
Their compact size makes them ideal for analytical applications, research environments, and sample preparation where space and precision are key. Made from high-grade borosilicate glass, they offer excellent chemical resistance and are reusable for sustainable lab workflows.
: In compliance with ISO 4796-80 and ASTM E128-99 standards.
Product Family Options
Product Information
- Plate ØPorosityCapacityMaterialHeightStem O-ØAvailabilityPrice
- 511-0384Micro-filter funnelMicro-filter funnel10 mm00,8 mlBorosilicate glass 3.320 mm6 mmLog in for availability1 * 5 ST114,00 €
- ROBU27081Micro-filter funnelMicro-filter funnel10 mm10,8 mlBorosilicate glass 3.320 mm6 mmLog in for availability1 * 5 ST114,00 €
- 511-0274Micro-filter funnelMicro-filter funnel10 mm20,8 mlBorosilicate glass 3.320 mm6 mmLog in for availability1 * 5 ST114,00 €
- 511-0256Micro-filter funnelMicro-filter funnel10 mm30,8 mlBorosilicate glass 3.320 mm6 mmLog in for availability1 * 5 ST114,00 €
- 511-0257Micro-filter funnelMicro-filter funnel10 mm40,8 mlBorosilicate glass 3.320 mm6 mmLog in for availability1 * 5 ST126,00 €
- 511-0385Micro-filter funnelMicro-filter funnel10 mm02 mlBorosilicate glass 3.345 mm6 mmLog in for availability1 * 5 ST114,00 €
- ROBU27201Micro-filter funnelMicro-filter funnel10 mm12 mlBorosilicate glass 3.345 mm6 mmLog in for availability1 * 5 ST114,00 €
- 511-0290Micro-filter funnelMicro-filter funnel10 mm22 mlBorosilicate glass 3.345 mm6 mmLog in for availability1 * 5 ST114,00 €
- 511-0291Micro-filter funnelMicro-filter funnel10 mm32 mlBorosilicate glass 3.345 mm6 mmLog in for availability1 * 5 ST114,00 €
- 511-0258Micro-filter funnelMicro-filter funnel10 mm42 mlBorosilicate glass 3.345 mm6 mmLog in for availability1 * 5 ST126,00 €