Funnels with short stem
Borosilicate glass 3.3.
- Good chemical and physical properties
- Not suitable for applications involving high thermal stress
- Quick and easy to clean
With smooth internal walls.
: DIN ISO 4798
Frequently Bought Together
Borosilicate glass 3.3.
With smooth internal walls.