DURAN® Funnels with short stem, ribbed
DURAN®, borosilicate glass 3.3, clear, ribbed walls.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For rapid filtration. Cone angle 60°. Funnel lip is ground and fire-polished.
Frequently Bought Together