Orders LinkContactUsLinkComponent
Funnels with long stem
Funnels with long stem
Supplier:   SciLabware
Funnels with long stem
Supplier:  SciLabware
Certificates
Funnels with long stem
Funnels with long stem
3 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls. Cone angle 60°.
Product Family Options
Product Information
  • Stem length
    Stem O-Ø
    For filter Ø
    Funnel O-Ø
    Availability
    Price
  • 150 mm
    8 mm
    110 - 125 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,00 €
    37,00 €
    1 * 1 ST
       37,00 €
  • 150 mm
    8 mm
    70 - 90 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,50 €
    25,50 €
    1 * 1 ST
       25,50 €
  • 150 mm
    8 mm
    125 - 150 mm
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,90 €
    24,90 €
    1 * 1 ST
       24,90 €