Orders LinkContactUsLinkComponent
Funnels with extra-long stem
Funnels with extra-long stem
Supplier:   KARTELL
Funnels with extra-long stem
Supplier:  KARTELL
Certificates
Funnels with extra-long stem
Funnels with extra-long stem
3 Options Available Below
Product Details & Documents
PP, transparent.
  • Inner ribs to accelerate the filtration process
  • Very good chemical resistance, autoclavable
  • Suitable for use with standard filter papers
With inner ribs for rapid filtration. Cone angle 60°.
Documents
Product Family Options
Product Information
  • Stem length
    Stem O-Ø
    Funnel O-Ø
    Availability
    Price
  • 150 mm
    8 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,30 €
    11,30 €
    1 * 1 ST
       11,30 €
  • 143 mm
    8 mm
    78,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,40 €
    12,40 €
    1 * 1 ST
       12,40 €
  • 145 mm
    8 mm
    103,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,80 €
    13,80 €
    1 * 1 ST
       13,80 €