Forceps for cover glasses
232-0111
- Type:Forceps for cover glasses, curved
- Length:105 mm
- Seq:01
- Pk:1
Steel, nickel-plated.
- For easy, secure handling of cover glasses
- Can also be used for other laboratory equipment, such as filter discs
- Easy to clean