Cover glass forceps
Supplier: HAMMACHER KARL
Jump to:
About this item
Wironit steel (18/12 stainless steel with 18% chrome and 10 - 12% nickel).
- For microscopic slides
- Resistant to corrosion
- Anti-magnetic
2 Options Available Below
Product Details & Documents
Wironit steel (18/12 stainless steel with 18% chrome and 10 - 12% nickel).
- For microscopic slides
- Resistant to corrosion
- Anti-magnetic