Orders LinkContactUsLinkComponent
Forceps
 :   BOCHEM
Forceps
 :  BOCHEM
Forceps
Forceps
Stainless steel, PTFE coating.
  • Anti-magnetic
  • Very high chemical and thermal resistance
  • Smooth surface prevents any contamination and makes cleaning easier
Product Family Options
  • Type
    Length
  • Blunt
    115 mm
    1 * 1 ST
    1 * 1 ST - 29,20 €
    29,20 €
    1 * 1 ST
       29,20 €
  • Blunt
    130 mm
    1 * 1 ST
    1 * 1 ST - 29,20 €
    29,20 €
    1 * 1 ST
       29,20 €
  • Blunt
    145 mm
    1 * 1 ST
    1 * 1 ST - 30,00 €
    30,00 €
    1 * 1 ST
       30,00 €