Order Entry
Austria
Orders LinkContactUsLinkComponent
Weighing forceps

Weighing forceps

Weighing forceps, 90 mm
Catalog # 232-0061
Supplier:   BOCHEM
Weighing forceps
Catalog # 232-0061
Supplier:  BOCHEM
Weighing forceps
Weighing forceps

Weighing forceps, 90 mm

Price
9,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, antimagnetic, with plastic tips.

  • Ideal for picking up and handling precision weights
  • Max. temperature 145 °C
Specifications
  • Description:
    Weighing forceps
  • Length:
    90 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.