Orders LinkContactUsLinkComponent
Forceps
Forceps
Supplier:   OUTILS RUBIS
Forceps
Supplier:  OUTILS RUBIS
Certificates
Forceps
5 Options Available Below
Product Details & Documents
Polybutene terephthalate (PBT) reinforced with 20% fibre glass, white. Universal forceps for a variety of laboratory and electronics-based applications.
  • Very good chemical resistance, anti-magnetic, high mechanical strength
  • High insulating values
  • Heat resistant up to 210 °C (for short periods)
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • 232-0210
    Straight, flat round tips
    Straight, flat round tips
    K2A
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,50 €
    15,50 €
    1 * 1 ST
       15,50 €
  • 232-1317
    Straight, flat wide tips
    Straight, flat wide tips
    K35A
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,50 €
    15,50 €
    1 * 1 ST
       15,50 €
  • 232-0204
    Straight, extra fine reinforced tips
    Straight, extra fine reinforced tips
    K7
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,50 €
    15,50 €
    1 * 1 ST
       15,50 €
  • 232-1319
    Straight, wide reinforced tips
    Straight, wide reinforced tips
    KR
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,50 €
    15,50 €
    1 * 1 ST
       15,50 €
  • 232-1313
    Curved, hook-shaped fine tips
    Curved, hook-shaped fine tips
    K6
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,50 €
    15,50 €
    1 * 1 ST
       15,50 €