Orders LinkContactUsLinkComponent
Flow indicators
Flow indicators
Supplier:   BURKLE
Flow indicators
Supplier:  BURKLE
Certificates
Flow indicators
6 Options Available Below
Product Details & Documents
These paddle wheel flow indicators give a clear visualisation of the flow rates of liquids or gas in tubing.
  • Colour coding - Red/Black/Blue
  • Flow indicator in either direction of flow and can be mounted at any angle
  • Paddle wheel impeller starts to rotate at about 150 ml/minute
LiquiMobil in SAN: Suitable for coolant circuits and contributes to reduction of water consumption appreciably. Maximum operating temperature 30 °C.
CheMobil in PMP: Chemical resistance makes for use with a great number of fluids. CheMobil is steam sterilisable. Maximum operating temperature 60 °C.
Acceptable maximum pressure 2 bar
Connector o.d.: 6 - 11 mm
Dimensions: 88×15×40 mm
Caution: LiquiMobil is not for use with aggressive liquids - CheMobil is not appropriate for strong acids, bases and solvents.
Product Family Options
Product Information
  • Type
    Colour
    Material
    Tube Ø
    Availability
    Price
  • 618-1111
    Flow indicator
    Flow indicator
    LiquiMobil
    Red
    SAN
    6 - 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,20 €
    11,20 €
    1 * 1 ST
       11,20 €
  • 618-1112
    Flow indicator
    Flow indicator
    LiquiMobil
    Black
    SAN
    6 - 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,20 €
    11,20 €
    1 * 1 ST
       11,20 €
  • 618-1110
    Flow indicator
    Flow indicator
    LiquiMobil
    Blue
    SAN
    6 - 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,20 €
    11,20 €
    1 * 1 ST
       11,20 €
  • 618-1121
    Flow indicator
    Flow indicator
    CheMobil
    Red
    PMP
    6 - 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,80 €
    13,80 €
    1 * 1 ST
       13,80 €
  • 618-1122
    Flow indicator
    Flow indicator
    CheMobil
    Black
    PMP
    6 - 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,80 €
    13,80 €
    1 * 1 ST
       13,80 €
  • 618-1120
    Flow indicator
    Flow indicator
    CheMobil
    Blue
    PMP
    6 - 11 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,80 €
    13,80 €
    1 * 1 ST
       13,80 €