Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Supplier:   Poulten & Graf
Volumetric flasks, class A
Supplier:  Poulten & Graf
Certificates
Volumetric flasks, class A
Volumetric flasks, class A
14 Options Available Below
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
 : DIN/BS/ISO 1042
Product Family Options
With works certificate
Product Information
  • Capacity
    NS
    Tolerance
    Availability
    Price
  • 5 ml
    10/13
    0,025 ± ml
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 64,80 €
    64,80 €
    1 * 5 ST
       64,80 €
  • 10 ml
    10/13
    0,025 ± ml
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 64,80 €
    64,80 €
    1 * 5 ST
       64,80 €
  • 20 ml
    10/19
    0,04 ± ml
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 67,10 €
    67,10 €
    1 * 5 ST
       67,10 €
  • 50 ml
    12/14
    0,06 ± ml
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 73,80 €
    73,80 €
    1 * 5 ST
       73,80 €
  • 200 ml
    14/15
    0,15 ± ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 41,40 €
    41,40 €
    1 * 2 ST
       41,40 €
  • 250 ml
    14/15
    0,15 ± ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 43,10 €
    43,10 €
    1 * 2 ST
       43,10 €
  • 500 ml
    19/17
    0,25 ± ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 51,30 €
    51,30 €
    1 * 2 ST
       51,30 €
Without certificate
Product Information
  • Capacity
    NS
    Tolerance
    Availability
    Price
  • 10 ml
    10/19
    0,025 ± ml
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 53,00 €
    53,00 €
    1 * 5 ST
       53,00 €
  • 20 ml
    10/13
    0,04 ± ml
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 55,40 €
    55,40 €
    1 * 5 ST
       55,40 €
  • 50 ml
    12/14
    0,06 ± ml
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 62,20 €
    62,20 €
    1 * 5 ST
       62,20 €
  • 200 ml
    14/15
    0,15 ± ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 36,70 €
    36,70 €
    1 * 2 ST
       36,70 €
  • 250 ml
    14/15
    0,15 ± ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 38,50 €
    38,50 €
    1 * 2 ST
       38,50 €
  • 500 ml
    19/17
    0,25 ± ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 46,70 €
    46,70 €
    1 * 2 ST
       46,70 €
  • 1000 ml
    24/20
    0,40 ± ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 73,70 €
    73,70 €
    1 * 2 ST
       73,70 €