Orders LinkContactUsLinkComponent
Volumetric flasks, class B
 :   Brand
Volumetric flasks, class B
 :  Brand
Volumetric flasks, class B
Volumetric flasks, class B
High clarity PP, with PP stopper.
  • Calibrated to contain (TC, In)
  • Ring mark and inscriptions in blue
  • Exposure to temperatures up to +60 °C (autoclaving) will not cause permanently exceeded error limits, Cleaning up to max. +60 °C is recommended to preserve marks and inscriptions
Error limits according to class B, DIN EN ISO 1042.
*Wide neck.
Product Family Options
  • Capacity
    NS
    Tolerance
  • 25 ml
    10/19
    0,08 ± ml
    1 * 1 ST
    1 * 1 ST - 10,10 €
    10,10 €
    1 * 1 ST
       10,10 €
  • 50 ml
    12/21
    0,12 ± ml
    1 * 1 ST
    1 * 1 ST - 13,00 €
    13,00 €
    1 * 1 ST
       13,00 €
  • 100 ml
    14/23
    0,2 ± ml
    1 * 1 ST
    1 * 1 ST - 13,90 €
    13,90 €
    1 * 1 ST
       13,90 €
  • 250 ml
    19/26
    0,3 ± ml
    1 * 1 ST
    1 * 1 ST - 17,10 €
    17,10 €
    1 * 1 ST
       17,10 €
  • 500 ml
    19/26
    0,5 ± ml
    1 * 1 ST
    1 * 1 ST - 17,90 €
    17,90 €
    1 * 1 ST
       17,90 €
  • 1000 ml
    24/29
    0,8 ± ml
    1 * 1 ST
    1 * 1 ST - 20,70 €
    20,70 €
    1 * 1 ST
       20,70 €
  • 10* ml
    10/19
    0,08
    1 * 1 ST
    1 * 1 ST - 9,90 €
    9,90 €
    1 * 1 ST
       9,90 €