Orders LinkContactUsLinkComponent
Volumetric flasks, class B
Volumetric flasks, class B
Supplier:   KARTELL
Volumetric flasks, class B
Supplier:  KARTELL
Certificates
Volumetric flasks, class B
Volumetric flasks, class B
5 Options Available Below
Product Details & Documents
PMP (TPX®), transparent, with stopper.
  • Autoclavable
  • For working temperatures up to +170 °C
  • With NS stoppers
Product Family Options
Product Information
  • Capacity
    NS
    Tolerance
    Availability
    Price
  • 50 ml
    14/23
    0,12 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,50 €
    23,50 €
    1 * 1 ST
       23,50 €
  • 100 ml
    14/23
    0,20 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,50 €
    26,50 €
    1 * 1 ST
       26,50 €
  • 250 ml
    19/26
    0,30 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • 500 ml
    19/26
    0,50 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,90 €
    37,90 €
    1 * 1 ST
       37,90 €
  • 1000 ml
    19/26
    0,80 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,70 €
    50,70 €
    1 * 1 ST
       50,70 €