Orders LinkContactUsLinkComponent
Volumetric flasks, class B, Nalgene®
   Sustainable Options Available
Volumetric flasks, class B, Nalgene®
   Sustainable Options Available
Certificates
Volumetric flasks, class B, Nalgene®
5 Options Available Below
Product Details & Documents
PMP (TPX®), transparent, with PP screw cap.
  • Very robust and allowing precise measurement
  • Calibrated individually with permanent colour graduation line
  • Autoclavable when the stopper has been removed from the threaded neck and a small quantity of water has been added to the flask. Cleaning with mild alkaline detergents recommended up to +60 °C (accuracy may be affected over +60 °C)
ASTM E288 and ISO Standard 384 Class B
 : ASTM E288 and ISO Standard 384 Class B
Specifications for Product Family
Specifications
  • Catalog No:
  • 612-3021
  • 612-3022
  • 612-3024
  • 612-3025
  • 612-3026
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Tolerance
    Availability
    Price
  • 50 ml
    0,10 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,80 €
    49,80 €
    1 * 1 ST
       49,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    612-3021
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 100 ml
    0,16 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,40 €
    53,40 €
    1 * 1 ST
       53,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    612-3022
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 250 ml
    0,24 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,70 €
    65,70 €
    1 * 1 ST
       65,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    612-3024
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 500 ml
    0,40 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 79,10 €
    79,10 €
    1 * 1 ST
       79,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    612-3025
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 1000 ml
    0,60 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 108,00 €
    108,00 €
    1 * 1 ST
       108,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    612-3026
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact