Orders LinkContactUsLinkComponent
Drying flasks
Drying flasks
Supplier:   BUCHI
Drying flasks
Supplier:  BUCHI
Certificates
Drying flasks
3 Options Available Below
Product Details & Documents
Made of Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For rotary evaporators Rotavapor R-210/215. Pear shape with NS 29/32 neck. Glassware A, V, C.
Product Family Options
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 500 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 151,00 €
    151,00 €
    1 * 1 ST
       151,00 €
  • 1000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 162,00 €
    162,00 €
    1 * 1 ST
       162,00 €
  • 2000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 230,00 €
    230,00 €
    1 * 1 ST
       230,00 €