ContactUsLinkComponent
Infusion bottles
Infusion bottles
Catalog # 215-9243
Supplier:  MUELLER KREMPEL
undefined
Infusion bottles
Catalog # 215-9243
Supplier:  MUELLER KREMPEL
Specifications
  • Capacity:
    1000 ml
  • Pk:
    1
Specifications
About this item
Clear glass, without closure.
  • Good chemical and physical properties
  • Screw caps and rubber inserts are available as an option
  • Double relief engraving and standard thread