Orders LinkContactUsLinkComponent
Flasks, pear shaped
Flasks, pear shaped
Supplier:   SciLabware
Flasks, pear shaped
Supplier:  SciLabware
Certificates
Flasks, pear shaped
Flasks, pear shaped
9 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for semi-micro distillations. With standard ground joint sleeve and white labelling field.
Product Family Options
Single Neck
Product Information
  • Capacity
    Socket size
    Centre neck
    Side necks
    Height
    Ext.Ø
    Availability
    Price
  • 25 ml
    14/23 NS
    100 mm
    38 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,40 €
    24,40 €
    1 * 1 ST
       24,40 €
  • 50 ml
    14/23 NS
    115 mm
    48 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,60 €
    26,60 €
    1 * 1 ST
       26,60 €
  • 100 ml
    14/23 NS
    135 mm
    58 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,60 €
    29,60 €
    1 * 1 ST
       29,60 €
  • 100 ml
    19/26 NS
    140 mm
    58 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,90 €
    30,90 €
    1 * 1 ST
       30,90 €
  • 10 ml
    14/23 NS
    85 mm
    23 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,70 €
    20,70 €
    1 * 1 ST
       20,70 €
  • 50 ml
    19/26 NS
    125 mm
    48 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,50 €
    31,50 €
    1 * 1 ST
       31,50 €
  • 100 ml
    24/29 NS
    150 mm
    58 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 155,00 €
    155,00 €
    1 * 5 ST
       155,00 €
  • 5 ml
    14/23 NS
    67 mm
    23 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 104,00 €
    104,00 €
    1 * 5 ST
       104,00 €
Two Necks
Product Information
  • Capacity
    Socket size
    Centre neck
    Side necks
    Height
    Ext.Ø
    Availability
    Price
  • 50 ml
    14/23 NS
    14/23 NS
    115 mm
    48 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,10 €
    56,10 €
    1 * 1 ST
       56,10 €