Orders LinkContactUsLinkComponent
Flasks, flat bottom with Quickfit® ground joints
Flasks, flat bottom with Quickfit® ground joints
Supplier:   SciLabware
Flasks, flat bottom with Quickfit® ground joints
Supplier:  SciLabware
Certificates
Flasks, flat bottom with Quickfit® ground joints
Flasks, flat bottom with Quickfit® ground joints
4 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, clear, with medium necks.
  • Very good chemical resistance
  • High temperature resistance
  • With longer neck than standard of flasks
Flat base allows use with hotplates. With labelling area.
Product Family Options
Product Information
  • Capacity
    Height
    Socket size
    Availability
    Price
  • 100 ml
    130 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 170,00 €
    170,00 €
    1 * 10 ST
       170,00 €
  • 50 ml
    150 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 40,60 €
    40,60 €
    1 * 2 ST
       40,60 €
  • 1000 ml
    190 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 324,00 €
    324,00 €
    1 * 10 ST
       324,00 €
  • 500 ml
    170 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 235,00 €
    235,00 €
    1 * 10 ST
       235,00 €