Orders LinkContactUsLinkComponent
Vacuum Filtering flasks, Büchner, with Quickfit® ground socket
Vacuum Filtering flasks, Büchner, with Quickfit® ground socket
Supplier:   SciLabware
Vacuum Filtering flasks, Büchner, with Quickfit® ground socket
Supplier:  SciLabware
Certificates
Vacuum Filtering flasks, Büchner, with Quickfit® ground socket
Vacuum Filtering flasks, Büchner, with Quickfit® ground socket
13 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Heavy wall and base construction for vacuum work
  • Side arm features a PTFE screwthread connector for the safe attachment of 9 mm bore vacuum tubing
Büchner flasks with precision-ground Quickfit® socket.
ISO 1773, BS 6352
Certifications: ISO 1773, BS 6352
Product Family Options
Plain glass side arm
Product Information
  • Capacity
    Height
    Ext.Ø
    Socket size
    Availability
    Price
  • 250 ml
    145 mm
    85 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 111,00 €
    111,00 €
    1 * 2 ST
       111,00 €
  • 250 ml
    145 mm
    85 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 617,00 €
    617,00 €
    1 * 10 ST
       617,00 €
  • 500 ml
    172 mm
    105 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 129,00 €
    129,00 €
    1 * 2 ST
       129,00 €
  • 500 ml
    172 mm
    105 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 658,00 €
    658,00 €
    1 * 10 ST
       658,00 €
  • 1000 ml
    227 mm
    136 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 170,00 €
    170,00 €
    1 * 2 ST
       170,00 €
  • 1000 ml
    227 mm
    136 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 813,00 €
    813,00 €
    1 * 10 ST
       813,00 €
PTFE screwthread tubing connector
Product Information
  • Capacity
    Height
    Ext.Ø
    Socket size
    Availability
    Price
  • 100 ml
    107 mm
    64 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 486,00 €
    486,00 €
    1 * 10 ST
       486,00 €
  • 100 ml
    107 mm
    64 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 471,00 €
    471,00 €
    1 * 10 ST
       471,00 €
  • 250 ml
    145 mm
    85 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 125,00 €
    125,00 €
    1 * 2 ST
       125,00 €
  • 250 ml
    145 mm
    85 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 617,00 €
    617,00 €
    1 * 10 ST
       617,00 €
  • 500 ml
    172 mm
    105 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 129,00 €
    129,00 €
    1 * 2 ST
       129,00 €
  • 1000 ml
    227 mm
    136 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 203,00 €
    203,00 €
    1 * 2 ST
       203,00 €
  • 1000 ml
    227 mm
    136 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 886,00 €
    886,00 €
    1 * 10 ST
       886,00 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.