Orders LinkContactUsLinkComponent
Flasks, Erlenmeyer, with standard ground joint
Flasks, Erlenmeyer, with standard ground joint
Supplier:   Lenz Laborglas
Flasks, Erlenmeyer, with standard ground joint
Supplier:  Lenz Laborglas
Certificates
Flasks, Erlenmeyer, with standard ground joint
Flasks, Erlenmeyer, with standard ground joint
36 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.
DIN 12387 (except Cat. No. 201-1114, 201-1122, 201-0318, 201-1116 and 271-1126)
Certifications: DIN 12387 (except Cat. No. 201-1114, 201-1122, 201-0318, 201-1116 and 271-1126)
Documents
Product Family Options
With socket NS 14/23
Product Information
  • Capacity
    Height
    Ext.Ø
    Socket size
    Availability
    Price
  • 10 ml
    60 mm
    42 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,80 €
    14,80 €
    1 * 1 ST
       14,80 €
  • 25 ml
    70 mm
    42 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,40 €
    15,40 €
    1 * 1 ST
       15,40 €
  • 50 ml
    85 mm
    51 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,60 €
    17,60 €
    1 * 1 ST
       17,60 €
  • 100 ml
    105 mm
    64 mm
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,10 €
    14,10 €
    1 * 1 ST
       14,10 €
With socket NS 19/26
Product Information
  • Capacity
    Height
    Ext.Ø
    Socket size
    Availability
    Price
  • 25 ml
    70 mm
    42 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,30 €
    11,30 €
    1 * 1 ST
       11,30 €
  • 50 ml
    85 mm
    51 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,30 €
    11,30 €
    1 * 1 ST
       11,30 €
  • 100 ml
    105 mm
    64 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,70 €
    13,70 €
    1 * 1 ST
       13,70 €
  • 150 ml
    131 mm
    74 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,20 €
    23,20 €
    1 * 1 ST
       23,20 €
  • 200 ml
    130 mm
    79 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,10 €
    20,10 €
    1 * 1 ST
       20,10 €
  • 250 ml
    140 mm
    85 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,90 €
    19,90 €
    1 * 1 ST
       19,90 €
  • 300 ml
    150 mm
    87 mm
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,70 €
    29,70 €
    1 * 1 ST
       29,70 €
With socket NS 24/29
Product Information
  • Capacity
    Height
    Ext.Ø
    Socket size
    Availability
    Price
  • 50 ml
    85 mm
    51 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,70 €
    12,70 €
    1 * 1 ST
       12,70 €
  • 100 ml
    105 mm
    64 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,70 €
    13,70 €
    1 * 1 ST
       13,70 €
  • 200 ml
    131 mm
    79 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,50 €
    19,50 €
    1 * 1 ST
       19,50 €
  • 250 ml
    140 mm
    85 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,20 €
    19,20 €
    1 * 1 ST
       19,20 €
  • 300 ml
    156 mm
    87 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,10 €
    20,10 €
    1 * 1 ST
       20,10 €
  • 500 ml
    175 mm
    105 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,10 €
    23,10 €
    1 * 1 ST
       23,10 €
  • 1000 ml
    215 mm
    131 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,10 €
    34,10 €
    1 * 1 ST
       34,10 €
  • 2000 ml
    280 mm
    166 mm
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,70 €
    66,70 €
    1 * 1 ST
       66,70 €
With socket NS 29/32
Product Information
  • Capacity
    Height
    Ext.Ø
    Socket size
    Availability
    Price
  • 50 ml
    85 mm
    51 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,20 €
    15,20 €
    1 * 1 ST
       15,20 €
  • 100 ml
    105 mm
    64 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,30 €
    12,30 €
    1 * 1 ST
       12,30 €
  • 150 ml
    131 mm
    74 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,80 €
    20,80 €
    1 * 1 ST
       20,80 €
  • 200 ml
    131 mm
    79 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,20 €
    17,20 €
    1 * 1 ST
       17,20 €
  • 250 ml
    140 mm
    85 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,80 €
    15,80 €
    1 * 1 ST
       15,80 €
  • 300 ml
    156 mm
    87 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,90 €
    17,90 €
    1 * 1 ST
       17,90 €
  • 500 ml
    175 mm
    105 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,10 €
    19,10 €
    1 * 1 ST
       19,10 €
  • 1000 ml
    220 mm
    131 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,20 €
    31,20 €
    1 * 1 ST
       31,20 €
  • 2000 ml
    280 mm
    166 mm
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,40 €
    62,40 €
    1 * 1 ST
       62,40 €
With socket NS 45/40
Product Information
  • Capacity
    Height
    Ext.Ø
    Socket size
    Availability
    Price
  • 200 ml
    131 mm
    79 mm
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,80 €
    24,80 €
    1 * 1 ST
       24,80 €
  • 250 ml
    140 mm
    85 mm
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,80 €
    24,80 €
    1 * 1 ST
       24,80 €
  • 300 ml
    159 mm
    87 mm
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,90 €
    28,90 €
    1 * 1 ST
       28,90 €
  • 500 ml
    175 mm
    105 mm
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,50 €
    39,50 €
    1 * 1 ST
       39,50 €
  • 1000 ml
    220 mm
    131 mm
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,40 €
    40,40 €
    1 * 1 ST
       40,40 €
  • 2000 ml
    280 mm
    166 mm
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 73,40 €
    73,40 €
    1 * 1 ST
       73,40 €
  • 3000 ml
    310 mm
    185 mm
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 134,00 €
    134,00 €
    1 * 1 ST
       134,00 €
  • 5000 ml
    365 mm
    223 mm
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 196,00 €
    196,00 €
    1 * 1 ST
       196,00 €