Orders LinkContactUsLinkComponent
Erlenmeyer flasks with baffles, Nalgene®
Erlenmeyer flasks with baffles, Nalgene®
   Sustainable Options Available
Erlenmeyer flasks with baffles, Nalgene®
   Sustainable Options Available
Certificates
Erlenmeyer flasks with baffles, Nalgene®
4 Options Available Below
Product Details & Documents
PC, transparent.
  • Thermal resistance from −135 to +135 °C
  • Shatter-proof with high mechanical strength
  • Good chemical resistance, but not suitable for solvents, strong acids or alkaline solutions
With four baffles seamlessly incorporated in the base. Ensures excellent mixing of liquids or microbiological cultures. Growth rates for E. coli and S. cerevisiae are comparable with glass flasks. The 500 and 1000 ml flasks have a straight neck accepting 38 mm standard caps made of rust-proof steel or plastic.
Specifications for Product Family
Specifications
  • Catalog No:
  • 710-4101
  • 710-4102
  • 710-4103
  • 710-4104
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 250 ml
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 91,20 €
    91,20 €
    1 * 4 ST
       91,20 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    710-4101
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 500 ml
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 111,00 €
    111,00 €
    1 * 4 ST
       111,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    710-4102
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 1000 ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 101,00 €
    101,00 €
    1 * 2 ST
       101,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    710-4103
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 2000 ml
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 129,00 €
    129,00 €
    1 * 2 ST
       129,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    710-4104
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact