Erlenmeyer flasks, disposable, with screw cap, Nalgene®
Lieferant: Thermo Fisher Scientific
Zertifikate
Über diesen Artikel
PETG, transparent, with HDPE screw cap.
- Leakproof HDPE screw cap closure can be opened partially for venting by twisting a quarter turn
- Packaged individually for easy storage and handling
- Reduced risk of cross-contamination
Sterile, disposable Erlenmeyer flasks reduce the risk of cross-contamination. Ideal for shakers and suspension cultures, and for creating or storing media. Raised graduations for approximate reading of the contents.
Spezifikationen
- Catalog No:
- 214-1925
- 214-0138
- 214-0137
- 214-0139
- 214-1926
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Spezifikationen
Zugehörige Informationen
Frequently Bought Together