ContactUsLinkComponent
Chemical burn spray, Diphoterine®
Chemical burn spray, Diphoterine®
Catalog # 118-1011
Supplier:  PREVOR EYEWASH
undefined
Chemical burn spray, Diphoterine®
Catalog # 118-1011
Supplier:  PREVOR EYEWASH
Specifications
  • Description:
    Chemical burn spray
  • Environmentally Preferable:
  • Pk:
    200 mL
Specifications
About this item
First aid procedures for chemical injuries often recommend rinsing the affected skin with water. But water causes the chemicals to penetrate deeper into the tissue. This first aid aerosol spray contains absorbing ingredients, which bind and neutralise chemicals immediately. The spray is sterile and has a shelf life of two years. 100 ml are needed to treat one cheek or hand, 200 ml for the face or one arm.
  • Absorbs and binds chemicals
  • Residues are neutral
  • Can be sprayed onto hands and face