Order Entry
Austria
ContactUsLinkComponent
 
Carbon Capsule Filter
  515-0000
undefined
Carbon Capsule Filter
  515-0000

 

  • Filtration area:
    70000 m² cm²
  • Max. temperature:
    80 °C
  • Inlet/outlet:
    3/8" FNPT; includes optional hose barb fittings to accept 12,7 mm ID tubing
  • Max. pressure:
    3,4 bar
  • Pk:
    1

 

 

Removes odor, colloids, and organics from liquids using activated carbon.

  • Finely divided carbon provides an active surface area of 65000 m²
  • Ready to use and disposable

Contains 100 g of acid-rinsed, activated charcoal. Inlet/outlet connections are ³/₈" FNPT. Includes optional hose barb adapters to accept ¹/₂" int.Ø tubing.

Applications:
- For laboratory and reagent-grade water and chemical filtration
- Protect laboratory from organic solvent odor