Syringe filters, Acrodisc®, Supor® membrane
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
Acrodisc™ syringe filters with Supor™ membrane deliver high flow rates and low protein binding.
- Superior flow rates and higher throughputs than competitive devices
- Low protein binding to minimize sample loss
- Pre-sterilized products are sterilized by gamma irradiation to eliminate potential contamination by EtO residuals
- A range of sizes (13 to 37 mm) accommodates sample volumes from < 10 to 150 ml
- Acrodisc PF and Serum Acrodisc syringe filters feature built-in prefilter for increased throughput of difficult-to-filter liquids
- Reduces mycoplasma with the use of 0,1 μm pore size
Supor™ hydrophilic polyethersulfone (PES) membrane: PP housing (except 25 and 32 mm: Acrylic and Acrodisc Serum: ABS)
Applications:
0,1 and 0,2 μm pore sizes provide sterilization of small volumes of buffers, culture media, and additives.
Acrodisc PF and Serum Acrodisc syringe filters are ideal for clarification/sterilization of viscous or particulate-laden solutions.
Use larger pore size filters for prefiltration and particulate removal.
Certifications: Passes United States Pharmacopeia (USP) Biological Reactivity Test, In Vivo <88>.
Specifications
- Catalog No:
- 514-4119
- 514-0861
- 514-4133
- 514-4134
- 514-4135
- 514-4122
- 514-4123
- 514-4129
- 514-4130
- 514-4131
- 514-0862
- 514-4132
- PALL4692
- 514-4102
- 514-1105
- 514-4124
- 514-4136
- 516-7654
- 517-0003
- 516-7701
- 516-7655
- 516-9087
- 514-4125
- 516-7703
- 514-4126
- 516-7656
- 514-4127
- 516-7657
- 514-4128
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Ø:
- 37 mm
- 25 mm
- 32 mm
- 32 mm
- 32 mm
- 13 mm
- 13 mm
- 32 mm
- 32 mm
- 32 mm
- 25 mm
- 32 mm
- 13 mm
- 25 mm
- 25 mm
- 13 mm
- 32 mm
- 25 mm
- 25 mm
- 32 mm
- 25 mm
- 32 mm
- 25 mm
- 32 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- Pore size:
- 0,20 µm
- 0,45 µm
- 0,45 µm
- 0,20 µm
- 1,20 µm
- 0,20 µm
- 0,45 µm
- 5,00 µm
- 0,10 µm
- 0,20 µm
- 0,45 µm
- 0,45 µm
- 0,20 µm
- 0,80/0,20 µm
- 0,20 µm
- 0,80 µm
- 0,80/0,20 µm
- 0,80/0,20 µm
- 0,10 µm
- 0,80/0,20 µm
- 0,20 µm
- 5,00 µm
- 0,10 µm
- 1,20/0,45 µm
- 0,20 µm
- 0,45 µm
- 0,45 µm
- 0,80 µm
- 0,80 µm
- Max. operating pressure:
- 5,2 bar
- 4,1 bar at 21 - 24 °C; 2,1 bar at 100 °C
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 4,1 bar at 21 - 24 °C; 2,1 bar at 100 °C
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 4,1 bar at 21 - 24 °C; 2,1 bar at 100 °C
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- 5,2 bar
- Max. temperature:
- 55 °C
- 100 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 100 °C
- 55 °C
- 55 °C
- 55 °C
- 100 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- Water flow rate:
- 425 ml/min (3,1 bar)
- 360 ml/min at 2,1 bar
- 700 ml/min (3,1 bar)
- 490 ml/min (3,1 bar)
- 1700 ml/min (3,1 bar)
- 22 ml/min (3,1 bar)
- 35 ml/min (3,1 bar)
- 1750 ml/min (3,1 bar)
- 100 ml/min (3,1 bar)
- 490 ml/min (3,1 bar)
- 360 ml/min at 2,1 bar
- 700 ml/min (3,1 bar)
- -
- 145 ml/min (3,1 bar)
- 360 ml/min at 2,1 bar
- 150 ml/min (3,1 bar)
- 440 ml/min (3,1 bar)
- 145 ml/min (3,1 bar)
- 35 ml/min (3,1 bar)
- 440 ml/min (3,1 bar)
- 175 ml/min (3,1 bar)
- 1750 ml/min (3,1 bar)
- 35 ml/min (3,1 bar)
- 1700 ml/min (3,1 bar)
- 175 ml/min (3,1 bar)
- 300 ml/min (3,1 bar)
- 300 ml/min (3,1 bar)
- 700 ml/min (3,1 bar)
- 700 ml/min (3,1 bar)
Specifications
Related Information
Frequently Bought Together